Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adck1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277296.1 | NP_001264225.1 | 4, 5 | 709 | ||
NM_001277297.1 | NP_001264226.1 | 4, 5 | 521 | |||
NM_028105.4 | NP_082381.1 | 4, 5 | 630 | |||
GenBank mRNA | AK011310.1 | - | 4, 5 | 481 | ||
AK050868.1 | - | 4, 5 | 611 | |||
AK154839.1 | - | 4, 5 | 576 | |||
AK210091.1 | - | 1, 2 | 38 | |||
BC010539.1 | - | 4, 5 | 523 | 4162365 |