Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005032.2 | - | 7 | 1955 | ||
AK152569.1 | - | 1 | 976 | |||
AK154976.1 | - | 8 | 2147 | |||
AK204721.1 | - | 1 | 282 | |||
BC050908.1 | - | 8 | 2130 | 6416955 | ||
BC053090.1 | - | 7 | 2215 | 5709246 |