Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myh14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271538.1 | NP_001258467.1 | 7 | 890 | ||
NM_001271540.1 | NP_001258469.1 | 9 | 955 | |||
NM_028021.3 | NP_082297.1 | 7 | 890 | |||
GenBank mRNA | AY205605.1 | - | 7 | 890 | ||
AY363100.1 | - | 9 | 955 | |||
EF602040.1 | - | 7 | 890 |