Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEFV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000243.2 | NP_000234.1 | 10 | 3006 | ||
NM_001198536.1 | NP_001185465.1 | 9 | 2548 | |||
GenBank mRNA | AF018080.1 | - | 10 | 3007 |