Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028038.3 | NP_082314.2 | 1 | 874 | ||
GenBank mRNA | AK010396.1 | - | 1 | 822 | ||
AK144945.1 | - | 1 | 1269 | |||
AK163480.1 | - | 1 | 827 | |||
BC058568.1 | - | 1 | 834 | |||
BC120556.1 | - | 1 | 874 | 40129479 | ||
BC120582.1 | - | 1 | 874 | 40129505 |