Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sulf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252578.1 | NP_001239507.1 | 19 | 2963 | ||
NM_001252579.1 | NP_001239508.1 | 19 | 2809 | |||
NM_028072.5 | NP_082348.2 | 19 | 2805 | |||
GenBank mRNA | AK008108.1 | - | 10 | 1068 | ||
AK028874.1 | - | 19 | 2720 | |||
AK034712.1 | - | 19 | 2790 | |||
AK036685.1 | - | 19 | 2741 | |||
AK049170.1 | - | 14 | 1720 | |||
AK081643.1 | - | 19 | 2763 | |||
AK129316.1 | - | 19 | 2963 | |||
AK133336.1 | - | 2779 | ||||
AK165183.1 | - | 19 | 2717 | |||
AY101177.1 | - | 18 | 2624 | |||
BC027238.1 | - | 8 | 779 | |||
BC062900.1 | - | 19 | 2761 | |||
BC141086.1 | - | 18 | 2558 | 9055698 |