Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Seh1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039088.1 | NP_001034177.1 | 5, 6 | 762 | ||
NM_028112.2 | NP_082388.1 | 5, 6 | 762 | |||
GenBank mRNA | AK011330.1 | - | 5, 6 | 664 | ||
AK088835.1 | - | 5, 6 | 636 | |||
AK143727.1 | - | 5, 6 | 762 | |||
AK166573.1 | - | 5, 6 | 627 | |||
AK169231.1 | - | 5, 6 | 643 | |||
AK169388.1 | - | 5, 6 | 642 | |||
AK170591.1 | - | 5, 6 | 715 | |||
BC027244.1 | - | 5, 6 | 714 | 3584697 |
Gene Symbol: 4930549G23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CN666923.1 | - | 2, 3 | 323 |