Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egln3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028133.2 | NP_082409.2 | 5 | 1317 | ||
GenBank mRNA | AF421882.1 | - | 5 | 1219 | ||
AJ310548.1 | - | 5 | 1298 | |||
AK011502.1 | - | 4 | 621 | |||
AK044787.1 | - | 5 | 1319 | |||
AK165972.1 | - | 5 | 1248 | |||
AK170732.1 | - | 5 | 1248 | |||
BC044926.1 | - | 5 | 1227 | 5373240 | ||
BC058278.1 | - | 5 | 1237 | 6528802 | ||
BC069893.1 | - | 5 | 1245 | 4978765 |