Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oraov1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028184.3 | NP_082460.2 | 2 | 157 | ||
GenBank mRNA | AK008702.1 | - | 2 | 217 | ||
AK046447.1 | - | 2 | 256 | |||
AK083244.1 | - | 2 | 214 | |||
AK167712.1 | - | 2 | 181 | |||
AK169310.1 | - | 2 | 181 | |||
BC006906.1 | - | 2 | 68 | |||
BC013564.1 | - | 2 | 157 | 3591291 |