Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028186.4 | NP_082462.3 | 4, 5 | 425 | ||
GenBank mRNA | AF358136.1 | - | 4, 5 | 375 | ||
AK008814.1 | - | 4, 5 | 407 | |||
AK081904.1 | - | 3, 4 | 371 | |||
AK146667.1 | - | 4, 5 | 407 | |||
AK158350.1 | - | 4, 5 | 407 | |||
AK165116.1 | - | 3, 4 | 371 | |||
AK199881.1 | - | 2, 3 | 87 | |||
AK203955.1 | - | 2, 3 | 88 |