Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Luc7l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025881.3 | NP_080157.1 | 2 | 209 | ||
NM_028190.3 | NP_082466.2 | 2 | 209 | |||
GenBank mRNA | AK007781.1 | - | 3 | 233 | ||
AK010539.1 | - | 2 | 161 | |||
AK017655.1 | - | 2 | 169 | |||
AK160293.1 | - | 2 | 210 | |||
BC019194.1 | - | 3 | 202 | |||
BC034135.1 | - | 1 | 375 | 5003664 | ||
BC055875.1 | - | 2 | 1192 | |||
BY726224.1 | - | 3 | 267 |