Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam192a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028221.4 | NP_082497.2 | 3 | 426 | ||
GenBank mRNA | AK010053.1 | - | 3 | 391 | ||
AK033988.1 | - | 3 | 357 | |||
AK050704.1 | - | 3 | 361 | |||
AK151603.1 | - | 3 | 380 | |||
BC016088.1 | - | 2 | 166 | 4503452 | ||
BC023158.1 | - | 3 | 265 | 4946201 | ||
BC025472.1 | - | 3 | 402 | 5353148 | ||
BC027060.1 | - | 3 | 388 | 5358022 | ||
BC027818.1 | - | 2 | 194 | 5320967 | ||
BC040818.1 | - | 379 | 4194531 |