Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem175 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163531.1 | NP_001157003.1 | 5 | 497 | ||
NM_001163532.1 | NP_001157004.1 | 5 | 500 | |||
NM_028223.3 | NP_082499.3 | 5 | 607 | |||
GenBank mRNA | AI526594.1 | - | 5 | 471 | ||
AK013879.1 | - | 5 | 430 | |||
AK034579.1 | - | 5 | 476 | |||
AK038865.1 | - | 5 | 467 | |||
AK049952.1 | - | 5 | 541 | |||
AK082765.1 | - | 5 | 463 | |||
AK170273.1 | - | 5 | 450 | |||
BB653267.1 | - | 5 | 537 | |||
BC005542.1 | - | 5 | 415 | 3590497 |