Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnmb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028231.2 | NP_082507.1 | 4 | 499 | ||
GenBank mRNA | AB193520.1 | - | 2 | 109 | ||
AK012400.1 | - | 4 | 350 | |||
AK014106.1 | - | 3 | 596 | |||
AY062429.1 | - | 2 | 109 | |||
BC046227.1 | - | 4 | 499 | 5703879 | ||
BC058957.1 | - | 4 | 499 | 5703879 |