Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114332.1 | NP_001107804.1 | 1 | 526 | ||
NM_028247.4 | NP_082523.1 | 1 | 526 | |||
GenBank mRNA | AK011813.1 | - | 1 | 530 | ||
AK036743.1 | - | 1 | 527 | |||
AK050229.1 | - | 1 | 529 | |||
AK144135.1 | - | 1 | 529 | |||
AK151290.1 | - | 1 | 527 | |||
AK151636.1 | - | 1 | 527 | |||
AK151704.1 | - | 1 | 531 | |||
AK157355.1 | - | 1 | 515 | |||
AK165240.1 | - | 1 | 527 | |||
AK165579.1 | - | 1 | 529 |