Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem55a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028264.4 | NP_082540.1 | 2 | 266 | ||
GenBank mRNA | AK012054.1 | - | 2 | 226 | ||
AK033845.1 | - | 2 | 229 | |||
AK049213.1 | - | 2 | 267 | |||
AK185686.1 | - | 2 | 78 | |||
AK206695.1 | - | 2 | 77 | |||
AX775959.1 | - | 2 | 195 | |||
BC021435.1 | - | 2 | 211 | 5029766 |