Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028303.3 | NP_082579.1 | 4, 5 | 341 | ||
GenBank mRNA | AK007454.1 | - | 4, 5 | 296 | ||
AK012624.1 | - | 4, 5 | 342 | |||
BC004608.1 | - | 4, 5 | 299 | 3585336 | ||
BC022181.1 | - | 3, 4 | 804 |