Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163746.1 | NP_001157218.1 | 2, 3 | 327 | ||
NM_028352.4 | NP_082628.3 | 2, 3 | 327 | |||
GenBank mRNA | AK013402.1 | - | 2, 3 | 310 | ||
AK028066.1 | - | 2, 3 | 317 | |||
AK032776.1 | - | 2, 3 | 279 | |||
AK036561.1 | - | 2, 3 | 313 | |||
AK043547.1 | - | 2, 3 | 328 | |||
AK049337.1 | - | 2, 3 | 279 | |||
AK049758.1 | - | 2, 3 | 314 | |||
AK051706.1 | - | 2, 3 | 267 | |||
AK153501.1 | - | 1, 2 | 147 | |||
AK160913.1 | - | 2, 3 | 328 | |||
AK165513.1 | - | 2, 3 | 312 | |||
AK169082.1 | - | 2, 3 | 292 | |||
AK169141.1 | - | 2, 3 | 286 | |||
AK169787.1 | - | 2, 3 | 301 | |||
BC138700.1 | - | 2, 3 | 252 | 8861722 |