Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028354.4 | NP_082630.2 | 2 | 179 | ||
GenBank mRNA | AK014855.1 | - | 2 | 160 | ||
AK033868.1 | - | 2 | 177 | |||
AK210567.1 | - | 2 | 379 | |||
BC065162.1 | - | 1 | 149 | 6402921 | ||
CJ151851.1 | - | 2 | 180 |