Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000264.3 | NP_000255.2 | 24 | 4753 | ||
NM_001083602.1 | NP_001077071.1 | 24 | 4861 | |||
NM_001083603.1 | NP_001077072.1 | 24 | 4707 | |||
NM_001083604.1 | NP_001077073.1 | 24 | 4471 | |||
NM_001083605.1 | NP_001077074.1 | 24 | 4454 | |||
NM_001083606.1 | NP_001077075.1 | 24 | 4553 | |||
NM_001083607.1 | NP_001077076.1 | 24 | 4627 | |||
GenBank mRNA | AB209495.1 | - | 21 | 7871 | ||
AK124593.1 | - | 3 | 1628 | |||
U43148.1 | - | 24 | 4553 |