Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chst14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028117.3 | NP_082393.3 | 1 | 973 | ||
GenBank mRNA | AK011230.1 | - | 1 | 910 | ||
AK154555.1 | - | 1 | 962 | |||
AK154636.1 | - | 1 | 942 | |||
AK155115.1 | - | 2 | 881 | |||
AK159357.1 | - | 1 | 941 | |||
AK171614.1 | - | 1 | 843 | |||
BC026886.1 | - | 1 | 794 | |||
BC043700.1 | - | 1 | 946 | 5346204 | ||
BC085479.1 | - | 1 | 890 | 6415106 |