Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc14a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171010.1 | NP_001164481.1 | 3 | 533 | ||
NM_001171011.1 | NP_001164482.1 | 2 | 268 | |||
NM_028122.4 | NP_082398.1 | 2 | 328 | |||
GenBank mRNA | AF448798.1 | - | 1 | 125 | ||
AJ420967.1 | - | 2 | 187 | |||
AK012066.1 | - | 2 | 178 | |||
AK041979.1 | - | 2 | 270 | |||
AK153891.1 | - | 3 | 438 | |||
BC058594.1 | - | 2 | 215 | 5364683 | ||
BC086673.1 | - | 2 | 224 | 30646772 | ||
BC100570.1 | - | 2 | 215 | 30044353 | ||
BY231473.1 | - | 2 | 328 |