Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msantd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145924.1 | NP_001139396.1 | 2 | 556 | ||
NM_001145925.1 | NP_001139397.1 | 2 | 417 | |||
NM_028137.3 | NP_082413.2 | 2 | 435 | |||
GenBank mRNA | AK011381.1 | - | 2 | 558 | ||
AK013199.1 | - | 2 | 436 | |||
AK017795.1 | - | 2 | 560 | |||
AK075956.1 | - | 2 | 429 | |||
BC052696.1 | - | 2 | 478 | 5715147 |