Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn7l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028139.5 | NP_082415.2 | 2 | 285 | ||
GenBank mRNA | AK013145.1 | - | 2 | 192 | ||
BB843098.1 | - | 2 | 208 | |||
BY203850.1 | - | 2 | 157 |