Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3gnt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028189.3 | NP_082465.3 | 3 | 1296 | ||
GenBank mRNA | AK008674.1 | - | 1335 | |||
AK144117.1 | - | 1286 | ||||
AK144180.1 | - | 3 | 1268 | |||
AY037785.1 | - | 1265 | ||||
AY039028.1 | - | 2 | 1215 | |||
BC026418.1 | - | 3 | 1288 | 4218953 |