Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289543.1 | NP_001276472.1 | 3, 4 | 646 | ||
NM_001289544.1 | NP_001276473.1 | 4, 5 | 970 | |||
NM_028227.3 | NP_082503.2 | 4, 5 | 842 | |||
GenBank mRNA | AF321920.1 | - | 4, 5 | 734 | ||
AF321921.1 | - | 4, 5 | 971 | |||
AK013885.1 | - | 2, 3 | 335 | |||
AK143718.1 | - | 3, 4 | 636 | |||
BC038490.1 | - | 1, 2 | 211 | 3599718 | ||
BC062932.1 | - | 4, 5 | 818 |