Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Immp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028260.2 | NP_082536.1 | 6 | 567 | ||
GenBank mRNA | AK005356.1 | - | 6 | 568 | ||
AK009570.1 | - | 6 | 525 | |||
AK010075.1 | - | 6 | 525 | |||
AK011278.1 | - | 6 | 568 | |||
AK011382.1 | - | 6 | 568 | |||
AK011420.1 | - | 6 | 529 | |||
AK011466.1 | - | 6 | 540 | |||
AK011618.1 | - | 6 | 529 | |||
AK011872.1 | - | 6 | 531 | |||
AK012181.1 | - | 520 | ||||
AK015978.1 | - | 6 | 538 | |||
AK078300.1 | - | 6 | 554 | |||
AK135670.1 | - | 6 | 527 | |||
AK168202.1 | - | 6 | 516 | |||
BC008259.1 | - | 6 | 526 | 2649406 | ||
BC081433.1 | - | 5 | 410 | 6814816 |