Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028262.3 | NP_082538.2 | 3 | 277 | ||
GenBank mRNA | AK011993.1 | - | 3 | 257 | ||
AK029403.1 | - | 3 | 257 | |||
AK031371.1 | - | 3 | 240 | |||
AK146777.1 | - | 3 | 271 | |||
AK166570.1 | - | 3 | 209 | |||
AY513271.1 | - | 3 | 210 | |||
BC016123.1 | - | 3 | 207 | 4503842 | ||
BC019973.1 | - | 3 | 207 | 4195130 | ||
BC057968.1 | - | 3 | 220 | 4979330 | ||
BC108358.1 | - | 3 | 248 | |||
BY087049.1 | - | 3 | 278 |