Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163746.1 | NP_001157218.1 | 4, 5 | 568 | ||
NM_028352.4 | NP_082628.3 | 4, 5 | 568 | |||
GenBank mRNA | AK013402.1 | - | 4, 5 | 551 | ||
AK028066.1 | - | 4, 5 | 558 | |||
AK032776.1 | - | 4, 5 | 520 | |||
AK036561.1 | - | 4, 5 | 554 | |||
AK043547.1 | - | 4, 5 | 569 | |||
AK049337.1 | - | 4, 5 | 520 | |||
AK049758.1 | - | 4, 5 | 555 | |||
AK051706.1 | - | 4, 5 | 508 | |||
AK153501.1 | - | 3, 4 | 388 | |||
AK160913.1 | - | 4, 5 | 569 | |||
AK165513.1 | - | 4, 5 | 553 | |||
AK169082.1 | - | 4, 5 | 533 | |||
AK169141.1 | - | 4, 5 | 527 | |||
AK169787.1 | - | 4, 5 | 542 | |||
BC138700.1 | - | 4, 5 | 493 | 8861722 |