Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDHA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000284.3 | NP_000275.1 | 10, 11 | 1161 | ||
NM_001173454.1 | NP_001166925.1 | 11, 12 | 1275 | |||
NM_001173455.1 | NP_001166926.1 | 10, 11 | 1182 | |||
NM_001173456.1 | NP_001166927.1 | 9, 10 | 1068 | |||
XM_017029574.1 | XP_016885063.1 | 10, 11 | 1169 | |||
GenBank mRNA | AK129887.1 | - | 10, 11 | 1096 | ||
AK222740.1 | - | 10, 11 | 1092 | |||
AK222988.1 | - | 10, 11 | 1126 | |||
AK293250.1 | - | 11, 12 | 1240 | |||
AK296341.1 | - | 10, 11 | 1148 | |||
AK296457.1 | - | 9, 10 | 982 | |||
AK312263.1 | - | 10, 11 | 1127 | |||
BC002406.1 | - | 10, 11 | 1073 | 2961286 | ||
EF576990.1 | - | 7, 8 | 630 | |||
EF590116.1 | - | 4, 5 | 811 | |||
EF590117.1 | - | 1193 | ||||
J03503.1 | - | 10, 11 | 1088 | |||
J03575.1 | - | 10, 11 | 1061 | |||
L13318.1 | - | 10, 11 | 1121 | |||
L48690.1 | - | 10, 11 | 1121 | |||
M24848.1 | - | 10, 11 | 1059 | |||
X52709.1 | - | 10, 11 | 1070 | |||
X52710.1 | - | 10, 11 | 1072 |