Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Faim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038658.2 | NP_001033747.1 | 12 | 1407 | ||
NM_028224.4 | NP_082500.2 | 12 | 1443 | |||
GenBank mRNA | AF468028.1 | - | 11 | 872 | ||
AK013476.1 | - | 12 | 846 | |||
AK030513.1 | - | 12 | 930 | |||
AK080839.1 | - | 12 | 957 | |||
AK081252.1 | - | 12 | 961 | |||
AK129246.1 | - | 12 | 956 | |||
AK144929.1 | - | 4 | 541 | |||
AK147343.1 | - | 8 | 8265 | |||
AK158960.1 | - | 12 | 1236 | |||
BC072560.1 | - | 12 | 1443 |