Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnmb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028231.2 | NP_082507.1 | 6 | 2446 | ||
GenBank mRNA | AK012400.1 | - | 6 | 2297 | ||
BC046227.1 | - | 6 | 2446 | 5703879 | ||
BC058957.1 | - | 6 | 2446 | 5703879 |