Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028238.7 | NP_082514.4 | 3 | 925 | ||
GenBank mRNA | AK009296.1 | - | 3 | 883 | ||
AK044783.1 | - | 3 | 888 | |||
AK054253.1 | - | 3 | 926 | |||
AK165682.1 | - | 3 | 928 | |||
AK199947.1 | - | 1 | 121 | |||
AK204142.1 | - | 1 | 228 | |||
AY062237.1 | - | 3 | 885 | |||
BC089578.1 | - | 3 | 885 | 6762755 |