Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028247.4 | NP_082523.1 | 3 | 1273 | ||
GenBank mRNA | AK050229.1 | - | 3 | 1276 | ||
AK165579.1 | - | 3 | 1276 |