Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028379.5 | NP_082655.1 | 7 | 950 | ||
GenBank mRNA | AK007572.1 | - | 5 | 935 | ||
AK013609.1 | - | 7 | 937 | |||
AK156167.1 | - | 7 | 947 | |||
AK177472.1 | - | 2 | 87 | |||
AK181896.1 | - | 2 | 82 | |||
BC019523.1 | - | 7 | 932 | 4163453 | ||
BC022770.1 | - | 1 | 24 |