Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pear1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032413.1 | NP_001027585.1 | 12, 13 | 1626 | ||
NM_001032414.1 | NP_001027586.1 | 11, 12 | 1556 | |||
NM_001289600.1 | NP_001276529.1 | 11, 12 | 1528 | |||
NM_001289601.1 | NP_001276530.1 | 13, 14 | 1769 | |||
NM_028460.2 | NP_082736.1 | 13, 14 | 1760 | |||
NM_152799.2 | NP_690012.2 | 13, 14 | 1725 | |||
GenBank mRNA | AF440279.1 | - | 13, 14 | 1750 | ||
AF444274.1 | - | 11, 12 | 1550 | |||
AF461685.1 | - | 10, 11 | 1483 | |||
AK053551.1 | - | 12, 13 | 1617 | |||
AK141082.1 | - | 1523 | ||||
AK161049.1 | - | 13, 14 | 1766 | |||
BC042490.1 | - | 13, 14 | 1485 | 4953584 |