Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028658.4 | NP_082934.3 | 2 | 221 | ||
GenBank mRNA | AK028185.1 | - | 2 | 85 | ||
AK155094.1 | - | 2 | 53 | |||
AK170254.1 | - | 2 | 109 | |||
BC106118.1 | - | 2 | 87 | |||
BC137673.1 | - | 2 | 161 | 8860692 | ||
BY205567.1 | - | 2 | 221 |