Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam98c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146023.1 | NP_001139495.1 | 7 | 916 | ||
GenBank mRNA | AK003496.1 | - | 6 | 840 | ||
AK080790.1 | - | 6 | 1008 | |||
AK138395.1 | - | 6 | 1819 | |||
AK166139.1 | - | 6 | 1815 | |||
AK171108.1 | - | 5 | 1282 | |||
BC023421.1 | - | 4 | 534 | 5029298 | ||
BC058765.1 | - | 6 | 973 |