Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028711.3 | NP_082987.1 | 2 | 169 | ||
GenBank mRNA | AB106930.1 | - | 2 | 313 | ||
AK014394.1 | - | 2 | 170 | |||
AK043831.1 | - | 2 | 369 | |||
AK054292.1 | - | 1 | 732 | |||
BC138993.1 | - | 2 | 176 | 8862015 | ||
BC138994.1 | - | 2 | 176 | 8862016 |