Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rap2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028712.4 | NP_082988.1 | 1 | 634 | ||
GenBank mRNA | AK014462.2 | - | 1 | 541 | ||
AK042941.1 | - | 1 | 526 | |||
AK045030.1 | - | 1 | 548 | |||
AK150187.1 | - | 1 | 518 | |||
AK163186.1 | - | 1 | 517 | |||
BC032168.1 | - | 1 | 614 | 5354704 | ||
BC046528.1 | - | 1 | 402 | 5149751 | ||
CT010292.1 | - | 1 | 84 |