Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014568.1 | - | 1, 2 | 482 | ||
AK020079.1 | - | 1, 2 | 64 | |||
AK042581.1 | - | 1, 2 | 468 | |||
AK046761.1 | - | 1, 2 | 473 | |||
AK047599.1 | - | 1, 2 | 497 | |||
AK050948.1 | - | 1, 2 | 480 | |||
AK079935.1 | - | 1, 2 | 480 | |||
AK157398.1 | - | 1, 2 | 480 | |||
AK157956.1 | - | 2, 3 | 385 | |||
AK164493.1 | - | 1, 2 | 491 | |||
AK172043.1 | - | 1, 2 | 486 | |||
BC024877.1 | - | 1, 2 | 484 | |||
BC051109.1 | - | 1, 2 | 453 | |||
BC096599.1 | - | 1, 2 | 472 | |||
BC125432.1 | - | 1, 2 | 479 | 40129947 | ||
BC132099.1 | - | 1, 2 | 479 | 40130376 |