Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271411.1 | NP_001258340.1 | 2 | 376 | ||
NM_001271412.1 | NP_001258341.1 | 2 | 306 | |||
NM_001271413.1 | NP_001258342.1 | 2 | 215 | |||
NM_001271414.1 | NP_001258343.1 | 2 | 303 | |||
NM_028728.3 | NP_083004.1 | 2 | 233 | |||
GenBank mRNA | AF361364.1 | - | 2 | 310 | ||
AK014802.2 | - | 2 | 234 | |||
AK048992.1 | - | 2 | 217 | |||
AK150635.1 | - | 2 | 285 | |||
AK151187.1 | - | 2 | 285 | |||
AK151884.1 | - | 2 | 285 | |||
AK152262.1 | - | 2 | 285 | |||
AK152499.1 | - | 2 | 285 | |||
AK152688.1 | - | 2 | 285 | |||
AK155106.1 | - | 2 | 230 | |||
AK162939.1 | - | 2 | 287 | |||
AK169918.1 | - | 2 | 230 | |||
AK170098.1 | - | 2 | 320 | |||
AK170355.1 | - | 2 | 283 | |||
AK170442.1 | - | 2 | 285 | |||
AK170545.1 | - | 2 | 171 | |||
AK170593.1 | - | 2 | 241 | |||
AK170677.1 | - | 2 | 241 | |||
AK170811.1 | - | 2 | 241 | |||
AK171082.1 | - | 2 | 346 | |||
AK171086.1 | - | 2 | 254 | |||
AK171201.1 | - | 2 | 215 | |||
BC150153.1 | - | 2 | 248 | |||
BC150488.1 | - | 2 | 248 | 40141021 |