Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hvcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042489.2 | NP_001035954.1 | 4 | 431 | ||
NM_028752.3 | NP_083028.1 | 3 | 293 | |||
GenBank mRNA | AK002854.1 | - | 3 | 294 | ||
AK154880.1 | - | 4 | 323 | |||
AK155123.1 | - | 4 | 268 | |||
AK170422.1 | - | 4 | 292 | |||
AK178408.1 | - | 2 | 155 | |||
BC021548.1 | - | 4 | 262 | 4913027 | ||
BY703352.1 | - | 3 | 287 | |||
CF734112.1 | - | 4 | 266 | |||
CF749152.1 | - | 4 | 431 | |||
CJ164124.1 | - | 4 | 429 |