Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kremen2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028416.2 | NP_082692.1 | 10 | 1705 | ||
GenBank mRNA | AJ457192.1 | - | 10 | 1717 | ||
AK013685.1 | - | 3 | 842 |