Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glrx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028419.2 | NP_082695.1 | 1 | 166 | ||
GenBank mRNA | AK009158.1 | - | 1 | 171 | ||
AK013761.1 | - | 1 | 167 | |||
AK050883.1 | - | 166 | ||||
BC050937.1 | - | 1 | 186 | |||
BC058371.1 | - | 1 | 186 | 6819353 | ||
DQ083330.1 | - | 1 | 155 |