Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glipr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028608.3 | NP_082884.1 | 2, 3 | 655 | ||
GenBank mRNA | AK010768.1 | - | 2, 3 | 587 | ||
AK137557.1 | - | 2, 3 | 602 | |||
AK170051.1 | - | 2, 3 | 602 | |||
BC025083.1 | - | 2, 3 | 579 | 3711757 | ||
BC098231.1 | - | 2, 3 | 629 | 3325823 |