Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011053.2 | NP_035183.2 | 36 | 5589 | ||
GenBank mRNA | AK003899.1 | - | 5 | 771 | ||
AK129080.1 | - | 36 | 5581 | |||
BC010844.1 | - | 36 | 5562 | |||
BC038503.1 | - | 7 | 1068 | |||
BC051231.1 | - | 4 | 916 | |||
BC055276.1 | - | 5 | 776 | |||
BC070468.1 | - | 36 | 5576 | 30628812 | ||
BC094244.1 | - | 16 | 2981 |