Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam98c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146023.1 | NP_001139495.1 | 7, 8 | 939 | ||
GenBank mRNA | AK003496.1 | - | 6, 7 | 863 | ||
AK080790.1 | - | 6, 7 | 1031 | |||
AK138395.1 | - | 6, 7 | 1842 | |||
AK166139.1 | - | 6, 7 | 1838 | |||
AK171108.1 | - | 5, 6 | 1305 | |||
BC023421.1 | - | 4, 5 | 557 | 5029298 | ||
BC058765.1 | - | 6, 7 | 996 |