Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028679.3 | NP_082955.2 | 5 | 606 | ||
GenBank mRNA | AF461763.1 | - | 5 | 586 | ||
AJ440757.2 | - | 5 | 528 | |||
AK014783.1 | - | 5 | 657 | |||
AK028436.1 | - | 519 | ||||
AK029057.1 | - | 5 | 607 | |||
AK049210.1 | - | 5 | 637 | |||
BC120829.1 | - | 5 | 567 | 40129752 | ||
BC120831.1 | - | 5 | 567 | 40129754 | ||
BC144835.1 | - | 5 | 567 |