Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028680.3 | NP_082956.2 | 3, 4 | 546 | ||
GenBank mRNA | AF274590.1 | - | 3, 4 | 564 | ||
AK014731.1 | - | 3, 4 | 598 | |||
AK047217.1 | - | 3, 4 | 546 | |||
BC138143.1 | - | 3, 4 | 520 | 8861162 | ||
BC138144.1 | - | 3, 4 | 520 | 8861163 | ||
DQ266091.1 | - | 3, 4 | 587 |